missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sol-M™ Standard Syringe with Fixed Hypodermic Needle
The Sol-M 3-Piece Syringe with Needle features low dead space technology that helps reduce medication waste and improve dosing accuracy.
Supplier: Sol-M™ 181025
Description
- High-transparency barrel to easily visualize contents
- Silicone lubrication ensures smooth plunger movement
- Plunger backstop that prevents accidental withdrawal of plunger from the barrel
- Large finger grips allow for easy, controlled injections
- Available in Luer Lock, Luer Slip Tip, and Catheter Tip options
Specifications
Not Autoclavable | |
25 G | |
5/8 in. | |
Blister | |
15.88 mm | |
Hypodermic Needle, Safety Needle, Blunt Fill Needle | |
Yes |
Polypropylene | |
Yes | |
Single-use | |
1 mL | |
Triple-bevel | |
Fixed | |
Sterile |