missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STAG2 Monoclonal Antibody (KT76), Invitrogen™
Rat Monoclonal Antibody
$1421.98 - $2134.69
Specifications
Antigen | STAG2 |
---|---|
Clone | KT76 |
Concentration | 1 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Description
STAG2 Monoclonal antibody specifically detects STAG2 in Human, Mouse samples. It is validated for Western BlotSpecifications
STAG2 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide | |
9230105L23Rik; B230112I07Rik; bA517O1.1; cohesin subunit SA-2; DKFZp686P168; DKFZp781H1753; FLJ25871; nuclear protein SA2; OTTHUMP00000024338; OTTHUMP00000043514; OTTHUMP00000043515; OTTHUMP00000081754; OTTHUMP00000081755; RGD1562042; RP11-517O1.1; SA2; SA-2; Sap2; SCC3 homolog 2; SCC3B; STAG2; Stromal antigen 2 | |
Recombinant mouse Stag2 (proprietary range information). | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
STAG2 |
KT76 | |
Western Blot | |
Unconjugated | |
Rat | |
Human, Mouse | |
O35638, Q8N3U4 | |
10735, 20843 | |
IgM | |
Protein L | |
Antibody |