Learn More
Ergodyne™ Squids™ 3405 Grabber with Belt Clip
Squids™ Grabbers are designed to hold a variety of objects like gloves, towels, and keys.
Supplier:  Ergodyne™ 19129
 View more versions of this product
Catalog No. 19-150-0007
Only null left
Explore available promotions
Description
Description
  • Slotted clip mounts to belt, PALS or like anchor
  • Durable, ultra-resilient acetal copolymer
  • Dielectric, non-conductive material
  • Belt clip mount keeps equipment close to the body
  • Breakaway feature
Specifications
Specifications
Belt Clip
Lime
Acetal Copolymer
Product Suggestions
Product Suggestions