missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Squalane 98.0+%, TCI America™
$35.41 - $227.41
Chemical Identifiers
CAS | 111-01-3 |
---|---|
Molecular Formula | C30H62 |
Molecular Weight (g/mol) | 422.826 |
MDL Number | MFCD00008953 |
InChI Key | PRAKJMSDJKAYCZ-UHFFFAOYSA-N |
Synonym | squalane, perhydrosqualene, dodecahydrosqualene, cosbiol, vitabiosol, spinacane, robane, hexamethyltetracosane, tetracosane, 2,6,10,15,19,23-hexamethyl, hexamethyl tetracosane |
PubChem CID | 8089 |
IUPAC Name | 2,6,10,15,19,23-hexamethyltetracosane |
SMILES | CC(C)CCCC(C)CCCC(C)CCCCC(C)CCCC(C)CCCC(C)C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
H009625ML
|
TCI America
H009625ML |
25 mL |
Each for $35.41
|
|
|||||
H0096100ML
|
TCI America
H0096100ML |
100 mL |
Each for $83.89
|
|
|||||
H0096500ML
|
TCI America
H0096500ML |
500 mL |
Each for $227.41
|
|
|||||
Chemical Identifiers
111-01-3 | |
422.826 | |
PRAKJMSDJKAYCZ-UHFFFAOYSA-N | |
8089 | |
CC(C)CCCC(C)CCCC(C)CCCCC(C)CCCC(C)CCCC(C)C |
C30H62 | |
MFCD00008953 | |
squalane, perhydrosqualene, dodecahydrosqualene, cosbiol, vitabiosol, spinacane, robane, hexamethyltetracosane, tetracosane, 2,6,10,15,19,23-hexamethyl, hexamethyl tetracosane | |
2,6,10,15,19,23-hexamethyltetracosane |
Specifications
111-01-3 | |
Colorless | |
C30H62 | |
25 mL | |
PRAKJMSDJKAYCZ-UHFFFAOYSA-N | |
2,6,10,15,19,23-hexamethyltetracosane | |
8089 | |
≥98.0% (GC) | |
Squalane |
-38°C | |
350°C | |
MFCD00008953 | |
squalane, perhydrosqualene, dodecahydrosqualene, cosbiol, vitabiosol, spinacane, robane, hexamethyltetracosane, tetracosane, 2,6,10,15,19,23-hexamethyl, hexamethyl tetracosane | |
CC(C)CCCC(C)CCCC(C)CCCCC(C)CCCC(C)CCCC(C)C | |
422.826 | |
422.83 | |
Liquid |
Safety and Handling
EINECSNumber : (9)-1317
RTECSNumber : XB6070000
TSCA : Yes