missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sorbitol, Powder, NF, 91-100.5%, Spectrum™ Chemical
SO219, 50-70-4, C6H14O6
$357.97 - $864.06
Chemical Identifiers
CAS | 50-70-4 |
---|---|
Molecular Formula | C6H14O6 |
Molecular Weight (g/mol) | 182.17 |
InChI Key | FBPFZTCFMRRESA-UHFFFAOYNA-N |
IUPAC Name | hexane-1,2,3,4,5,6-hexol |
SMILES | OCC(O)C(O)C(O)C(O)CO |
Description
Spectrum™ Chemical Sorbitol, Powder, NF is used in the pharmaceutical industry as a sweetener or humectant. Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
50-70-4 | |
182.17 | |
hexane-1,2,3,4,5,6-hexol |
C6H14O6 | |
FBPFZTCFMRRESA-UHFFFAOYNA-N | |
OCC(O)C(O)C(O)C(O)CO |
Specifications
50-70-4 | |
3.5 to 7.0 | |
Poly Bottle | |
500 g | |
OCC(O)C(O)C(O)C(O)CO | |
182.17 | |
NF |
1 | |
0.001 | |
C6H14O6 | |
FBPFZTCFMRRESA-UHFFFAOYNA-N | |
hexane-1,2,3,4,5,6-hexol | |
91 to 100.5% |