Learn More
ABS Solid Door Laboratory Refrigerator/Freezer Combo Auto Defrost
Solid door Laboratory refrigerator/freezer combo units meet and exceed the specifications of most general-purpose units, providing both refrigerator and freezer compartments with standard analog dial temperature thermostats.
Supplier:  ABS ABTHCRFC20A
Catalog No. 361058585
Only null left
Explore available promotions
Description
Description
Units are intended for general purpose applications only where precise temperature control and maintenance are not essential and there are no requirements for temperature alarms.
  • Auto defrost freezer compartments have programmed defrost cycles that will reach air temperatures of approximately 0 degrees Celsius.
  • Environmentally friendly, natural hydrocarbon refrigerants (not containing hydrofluorocarbons) vastly reduce global-warming potential (GWP) while meeting new EPA/SNAP mandates and UL, ASHRAE compliance guidelines.
Specifications
Specifications
Laboratory Refrigerator/Freezer
566 L
Solid Swing
3
Steel
Mechanical Thermostat
NEMA 5-15
20 cu. ft.
0°C to 10°C
2
Steel
Upright
CFC-free
115 V
Product Suggestions
Product Suggestions