missing translation for 'onlineSavingsMsg'
Learn More

Sodium Trifluoromethanesulfinate 95.0+%, TCI America™

$151.17 - $476.22

Chemical Identifiers

CAS 2926-29-6
Molecular Formula CF3NaO2S
Molecular Weight (g/mol) 156.054
MDL Number MFCD03092989
InChI Key KAVUKAXLXGRUCD-UHFFFAOYSA-M
Synonym sodium trifluoromethanesulfinate, sodium trifluoromethanesulphinate, sodium trifluoro-methanesulfinate, trifluoromethanesulfinic acid sodium salt, methanesulfinic acid, trifluoro-, sodium salt, langlois reagent, chf3o2s.na, acmc-209h7v, ksc563a2r, sodium trifluoromethylsulfinate
PubChem CID 23690734
IUPAC Name sodium;trifluoromethanesulfinate
SMILES C(F)(F)(F)S(=O)[O-].[Na+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
T20335G
View Documents
TCI America
T20335G
5 g N/A
Only null left
 
T203325G
View Documents
TCI America
T203325G
25 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

2926-29-6
156.054
KAVUKAXLXGRUCD-UHFFFAOYSA-M
23690734
C(F)(F)(F)S(=O)[O-].[Na+]
CF3NaO2S
MFCD03092989
sodium trifluoromethanesulfinate, sodium trifluoromethanesulphinate, sodium trifluoro-methanesulfinate, trifluoromethanesulfinic acid sodium salt, methanesulfinic acid, trifluoro-, sodium salt, langlois reagent, chf3o2s.na, acmc-209h7v, ksc563a2r, sodium trifluoromethylsulfinate
sodium;trifluoromethanesulfinate

Specifications

2926-29-6
CF3NaO2S
5 g
KAVUKAXLXGRUCD-UHFFFAOYSA-M
sodium;trifluoromethanesulfinate
23690734
≥95.0% (T)
Sodium Trifluoromethanesulfinate
White-Yellow
MFCD03092989
sodium trifluoromethanesulfinate, sodium trifluoromethanesulphinate, sodium trifluoro-methanesulfinate, trifluoromethanesulfinic acid sodium salt, methanesulfinic acid, trifluoro-, sodium salt, langlois reagent, chf3o2s.na, acmc-209h7v, ksc563a2r, sodium trifluoromethylsulfinate
C(F)(F)(F)S(=O)[O-].[Na+]
156.054
156.05
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'