missing translation for 'onlineSavingsMsg'
Learn More

Sodium Thiosulfate, Certified, 0.1000N ±0.0005N (0.1M), LabChem™

$34.13 - $244.90

Chemical Identifiers

CAS 10102-17-7
Molecular Formula H10Na2O8S2
Molecular Weight (g/mol) 248.17
InChI Key PODWXQQNRWNDGD-UHFFFAOYSA-L
Synonym sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate
PubChem CID 61475
ChEBI CHEBI:32150
IUPAC Name disodium pentahydrate sulfanidesulfonate
SMILES O.O.O.O.O.[Na+].[Na+].[O-]S([S-])(=O)=O
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
LC250601
View Documents
LabChem
LC250601
500 mL
Each for $34.13
Only null left
 
LC250602
View Documents
LabChem
LC250602
1 L
Each for $42.77
Only null left
 
LC250604
View Documents
LabChem
LC250604
4 L
Each for $96.27
Only null left
 
LC250606
View Documents
LabChem
LC250606
10 L
Each for $181.23
Only null left
 
LC250605
View Documents
LabChem
LC250605
20 L
Each for $244.90
Only null left
 
Description

Description

Specifications

Chemical Identifiers

10102-17-7
248.17
sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate
CHEBI:32150
O.O.O.O.O.[Na+].[Na+].[O-]S([S-])(=O)=O
H10Na2O8S2
PODWXQQNRWNDGD-UHFFFAOYSA-L
61475
disodium pentahydrate sulfanidesulfonate

Specifications

10102-17-7,7732-18-5,497-19-8
2.48,97.5,0.02
500 mL
Na2S2O3·5H2O
Soluble in water
O.O.O.O.O.[Na+].[Na+].[O-]S([S-])(=O)=O
248.17
CHEBI:32150
Certified
Passes Test
Sulfur compounds; Carbon dioxide
Colorless
Liquid
H10Na2O8S2
sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate
PODWXQQNRWNDGD-UHFFFAOYSA-L
disodium pentahydrate sulfanidesulfonate
61475
248.18
0.1000N ±0.0005N (0.1M)
Poly Bottle
Sodium Thiosulfate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Solution is not hazardous.

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

Recommended Storage : Room Temperature

SDS
missing translation for 'documents'

missing translation for 'documents'