missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Tartrate, Dihydrate, Crystal, Reagent, ACS, 99-101%, Spectrum™ Chemical
S1485, 6106-24-7, Na2C4H4O6·2H2O
Supplier: Spectrum Chemical Mfg Cor S148512KGBL
Description
Spectrum™ Chemical Sodium Tartrate, Dihydrate, Crystal, Reagent, ACS has a crystal structure that is able to capture a very precise amount of water and is used in the Karl Fischer moisture determination titration technique. As an ACS grade reagent, Spectrum Chemical manufactured compound is used as the quality standard against which other substances are grade and has met the toughest regulatory standards for quality and pureness.Specifications
6106-24-7 | |
7.0 to 9.0 | |
Poly Pail | |
MFCD00150035 | |
FGJLAJMGHXGFDE-UDMGOBQRNA-L | |
disodium (2R,3R)-2,3-dihydroxybutanedioate dihydrate | |
99.0 to 101% |
1 | |
15.61-15.71% | |
C4H8Na2O8 | |
12 kg | |
O.O.[Na+].[Na+].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O | |
230.08 | |
ACS |
Chemical Identifiers
6106-24-7 | |
230.08 | |
FGJLAJMGHXGFDE-UDMGOBQRNA-L | |
O.O.[Na+].[Na+].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O |
C4H8Na2O8 | |
MFCD00150035 | |
disodium (2R,3R)-2,3-dihydroxybutanedioate dihydrate |