missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydroxide Solution, 1 M, For HPCE, MilliporeSigma™ Supelco™

$170.72 - $553.33

Chemical Identifiers

MDL Number MFCD00003548
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
111019410
View Documents
Merck Emd Millipore
79724100ML
100 mL
Each for $170.72
Only null left
 
111016646
View Documents
Merck Emd Millipore
79724500ML
500 mL
Each for $553.33
Only null left
 
Description

Description

Specifications

Chemical Identifiers

MFCD00003548

Specifications

1310-73-2
MFCD00003548
40.00
0.9-1.1 M
NaOH
100 mL
HPCE
3 mmHg (37°C)
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'