missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide, 50% w/w Reagent (760 g/L), Reagents
Sodium Hydroxide, 50% w/w Reagent (760 g/L), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc C13186001A
Specifications
>13 | |
Colorless | |
7732-18-5,1310-73-2 | |
H2O | |
1 L | |
O | |
18.02 | |
Poly Bottle | |
Liquid |
Sodium Hydroxide, 50% w/w Reagent (760 g/L) | |
140°C | |
12°C | |
Lye, Caustic soda | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
water | |
Reagent | |
1.5267 |
Chemical Identifiers
7732-18-5 | |
18.02 | |
Lye, Caustic soda | |
O |
H2O | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
water |