missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 10.0 N, from NF/USP material, Ricca Chemical

$417.63 - $417.63

Chemical Identifiers

CAS 1310-73-2
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
R74700504A
SDS View Documents
Ricca Chemical Company
R74700504A
4 L
Each for $417.63
Only null left
 
Description

Description

Specifications

Chemical Identifiers

1310-73-2
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
14798
CHEBI:32145

Specifications

Colorless
70.0,28.42
HNaO
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium;hydroxide
14798
Laboratory
Liquid
1310-73-2,7732-18-5
72.0,29.58
10.0Normal
4 L
[OH-].[Na+]
39.997
CHEBI:32145
Natural Poly Bottle
Sodium Hydroxide Solution
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'