missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 0.600 N, Ricca Chemical

$104.59 - $104.59

Chemical Identifiers

CAS 1310-73-2
Molecular Weight (g/mol) 40 g/mol
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
7416-32
SDS View Documents
Ricca Chemical Company
741632
1 L Natural Poly Bottle
Each for $104.59
Only null left
 
Description

Description

  • Tested in an ISO 17025 Accredited Facility
  • Test Reports provided with Certified Values and Measurement Uncertainty
  • Tight release specifications for low lot-to-lot variability
Specifications

Chemical Identifiers

1310-73-2
40 g/mol

Specifications

1310-73-2
40 g/mol
0.600N
Sodium Hydroxide
1 L
Laboratory
Natural Poly Bottle
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'