missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 0.500 N (N/2), Ricca Chemical

$68.66 - $476.69

Chemical Identifiers

Molecular Formula HNaO
Molecular Weight (g/mol) 40 g/mol
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
740016
SDS Accessories View Documents
Ricca Chemical Company
740016
500 mL Natural Poly Bottle
Each for $68.66
Only null left
 
7400-32
SDS Accessories View Documents
Ricca Chemical Company
740032
1 L Natural Poly Bottle
Each for $80.66
Only null left
 
7400-1
SDS Accessories View Documents
Ricca Chemical Company
74001
4 L Natural Poly Bottle
Each for $185.06
Only null left
 
74002-5
View Documents
Ricca Chemical Company
740025
10 L Cubitainer
Each for $353.97
Only null left
 
7400-5
View Documents
Ricca Chemical Company
74005
20 L Cubitainer
Each for $476.69
Only null left
 
Description

Description

  • Tested in an ISO 17025 Accredited Facility
  • Test Reports provided with Certified Values and Measurement Uncertainty
  • Tight release specifications for low lot-to-lot variability
Specifications

Chemical Identifiers

HNaO
HEMHJVSKTPXQMS-UHFFFAOYSA-M
14798
sodium;hydroxide
40 g/mol
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
CHEBI:32145
[OH-].[Na+]

Specifications

Colorless
98
0.500 N
500 mL
[OH-].[Na+]
40 g/mol
CHEBI:32145
Natural Poly Bottle
Sodium Hydroxide
94
HNaO
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium;hydroxide
14798
Laboratory
Liquid
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'