missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 0.05N Volumetric Solution, BAKER ANALYZED™ Reagent, J.T. Baker™

High quality chemicals for laboratory and specialized industrial use

$2761.00 - $3980.00

Chemical Identifiers

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
02004106
View Documents
Avantor
566402
1 L Narrow Mouth Polyethylene Bottle
Case of 6 Each for $2,761.00
Only null left
 
02004107
View Documents
Avantor
566403
4 L Cubetainer
Case of 4 Each for $3,980.00
Only null left
 
Description

Description

Specifications

Chemical Identifiers

1310-73-2
39.997
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
CHEBI:32145
[OH-].[Na+]
HNaO
HEMHJVSKTPXQMS-UHFFFAOYSA-M
14798
sodium;hydroxide

Specifications

1310-73-2
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium;hydroxide
14798
40
Narrow Mouth Polyethylene Bottle
Sodium Hydroxide, 0.05N Volumetric Solution
HNaO
1 L
[OH-].[Na+]
39.997
CHEBI:32145
BAKER ANALYZED™ Reagent
1L = 1.00kg
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'