missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Dodecyl Sulfate 97.0+%, TCI America™
$61.51 - $277.75
Chemical Identifiers
CAS | 151-21-3 |
---|---|
Molecular Formula | C12H25NaO4S |
Molecular Weight (g/mol) | 288.38 |
MDL Number | MFCD00036175 |
InChI Key | DBMJMQXJHONAFJ-UHFFFAOYSA-M |
Synonym | sodium dodecyl sulfate, sodium lauryl sulfate, sodium dodecylsulfate, sodium lauryl sulphate, sodium dodecyl sulphate, dodecyl sulfate, sodium salt, anticerumen, gardinol, neutrazyme, duponal |
PubChem CID | 3423265 |
ChEBI | CHEBI:8984 |
IUPAC Name | sodium dodecyl sulfate |
SMILES | [Na+].CCCCCCCCCCCCOS([O-])(=O)=O |
Chemical Identifiers
151-21-3 | |
288.38 | |
DBMJMQXJHONAFJ-UHFFFAOYSA-M | |
3423265 | |
sodium dodecyl sulfate |
C12H25NaO4S | |
MFCD00036175 | |
sodium dodecyl sulfate, sodium lauryl sulfate, sodium dodecylsulfate, sodium lauryl sulphate, sodium dodecyl sulphate, dodecyl sulfate, sodium salt, anticerumen, gardinol, neutrazyme, duponal | |
CHEBI:8984 | |
[Na+].CCCCCCCCCCCCOS([O-])(=O)=O |
Specifications
151-21-3 | |
White | |
MFCD00036175 | |
sodium dodecyl sulfate, sodium lauryl sulfate, sodium dodecylsulfate, sodium lauryl sulphate, sodium dodecyl sulphate, dodecyl sulfate, sodium salt, anticerumen, gardinol, neutrazyme, duponal | |
[Na+].CCCCCCCCCCCCOS([O-])(=O)=O | |
288.38 | |
CHEBI:8984 | |
≥97.0% (T) | |
Sodium Dodecyl Sulfate [Reagent for Ion-Pair Chromatography] |
207°C | |
C12H25NaO4S | |
25 g | |
DBMJMQXJHONAFJ-UHFFFAOYSA-M | |
sodium dodecyl sulfate | |
3423265 | |
288.38 | |
Crystalline Powder |
Safety and Handling
EINECSNumber : (2)-1679
RTECSNumber : WT1050000
TSCA : Yes