missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Chloride, ACS Reagent Grade, Ricca Chemical
$165.44 - $165.44
Chemical Identifiers
CAS | 7647-14-5 |
---|---|
Molecular Formula | ClNa |
Molecular Weight (g/mol) | 58.44 |
MDL Number | MFCD00003477 |
InChI Key | FAPWRFPIFSIZLT-UHFFFAOYSA-M |
ChEBI | CHEBI:26710 |
IUPAC Name | sodium chloride |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
RDCS0430500
|
Ricca Chemical Company
RDCS0430500B1 |
500 g |
Each for $165.44
|
|
|||||
Chemical Identifiers
7647-14-5 | |
58.44 | |
FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
sodium chloride |
ClNa | |
MFCD00003477 | |
CHEBI:26710 |
Specifications
White | |
98.0 | |
Powder | |
ClNa | |
sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex | |
[Na+].[Cl-] | |
58.44 | |
CHEBI:26710 | |
ACS | |
Sodium Chloride |
7647-14-5 | |
100.0 | |
500 g | |
MFCD00003477 | |
FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
sodium chloride | |
5234 | |
≥ 99% | |
White Poly Wide Mouth Bottle |