missing translation for 'onlineSavingsMsg'
Learn More

Sodium Bis(trifluoromethanesulfonyl)imide 98.0+%, TCI America™

$160.63 - $480.00

Chemical Identifiers

CAS 91742-21-1
Molecular Formula C2HF6NNaO4S2
Molecular Weight (g/mol) 304.133
MDL Number MFCD14156021
InChI Key QXZNUMVOKMLCEX-UHFFFAOYSA-N
Synonym Bis(trifluoromethanesulfonyl)imide Sodium Salt, Sodium Triflimide
PubChem CID 131887538
IUPAC Name sodium;1,1,1-trifluoro-N-(trifluoromethylsulfonyl)methanesulfonamide
SMILES C(F)(F)(F)S(=O)(=O)NS(=O)(=O)C(F)(F)F.[Na]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
S09891G
View Documents
TCI America
S09891G
1 g
Each for $160.63
Only null left
 
S09895G
View Documents
TCI America
S09895G
5 g
Each for $480.00
Only null left
 
Description

Description

Specifications

Chemical Identifiers

91742-21-1
304.133
QXZNUMVOKMLCEX-UHFFFAOYSA-N
131887538
C(F)(F)(F)S(=O)(=O)NS(=O)(=O)C(F)(F)F.[Na]
C2HF6NNaO4S2
MFCD14156021
Bis(trifluoromethanesulfonyl)imide Sodium Salt, Sodium Triflimide
sodium;1,1,1-trifluoro-N-(trifluoromethylsulfonyl)methanesulfonamide

Specifications

91742-21-1
White
MFCD14156021
1759
QXZNUMVOKMLCEX-UHFFFAOYSA-N
sodium;1,1,1-trifluoro-N-(trifluoromethylsulfonyl)methanesulfonamide
131887538
≥98.0% (T)
Sodium Bis(trifluoromethanesulfonyl)imide
257°C
C2HF6NNaO4S2
1 g
Bis(trifluoromethanesulfonyl)imide Sodium Salt, Sodium Triflimide
C(F)(F)(F)S(=O)(=O)NS(=O)(=O)C(F)(F)F.[Na]
304.133
303.13
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents