missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Bis(fluorosulfonyl)imide 98.0+%, TCI America™
$334.49 - $941.11
Chemical Identifiers
CAS | 100669-96-3 |
---|---|
Molecular Formula | F2NNaO4S2 |
Molecular Weight (g/mol) | 203.11 |
InChI Key | VCCATSJUUVERFU-UHFFFAOYSA-N |
PubChem CID | 102310776 |
IUPAC Name | sodium;bis(fluorosulfonyl)azanide |
SMILES | [N-](S(=O)(=O)F)S(=O)(=O)F.[Na+] |
Chemical Identifiers
100669-96-3 | |
203.11 | |
102310776 | |
[N-](S(=O)(=O)F)S(=O)(=O)F.[Na+] |
F2NNaO4S2 | |
VCCATSJUUVERFU-UHFFFAOYSA-N | |
sodium;bis(fluorosulfonyl)azanide |
Specifications
100669-96-3 | |
White | |
1 g | |
VCCATSJUUVERFU-UHFFFAOYSA-N | |
sodium;bis(fluorosulfonyl)azanide | |
102310776 | |
≥98.0% (T) | |
Sodium Bis(fluorosulfonyl)imide |
112°C | |
F2NNaO4S2 | |
1759 | |
[N-](S(=O)(=O)F)S(=O)(=O)F.[Na+] | |
203.11 | |
203.11 | |
Crystalline Powder |
Safety and Handling
TSCA : No