missing translation for 'onlineSavingsMsg'
Learn More

Sodium Acetate, Anhydrous, USP, 99-101%, Spectrum™ Chemical

SO104, 127-09-3, C2H3NaO2

Supplier:  Spectrum Chemical Mfg Cor SO10412KG

 View more versions of this product

Catalog No. 18606267


Only null left
Explore available promotions

Description

Description

Spectrum™ Chemical Sodium Acetate, Anhydrous, USP is used as an electrolyte replenisher in isosmotic solution for parenteral replacement of acute losses of extracellular fluid without disturbing normal electrolyte balance. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Note: This item is non-returnable.

 

Specifications

Specifications

7.5 to 9.2
1
MFCD00012459
[Na+].CC([O-])=O
82.03
USP
0.01
127-09-3
C2H3NaO2
VMHLLURERBWHNL-UHFFFAOYSA-M
sodium acetate
99 to 101%
12 kg
Poly Pail

Chemical Identifiers

127-09-3
82.03
VMHLLURERBWHNL-UHFFFAOYSA-M
[Na+].CC([O-])=O
C2H3NaO2
MFCD00012459
sodium acetate
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions