missing translation for 'onlineSavingsMsg'
Learn More

Sodium 4-Hydroxybenzenesulfonate Dihydrate 98.0+%, TCI America™

$33.56 - $160.30

Chemical Identifiers

CAS 10580-19-5
Molecular Formula C6H5O4S
Molecular Weight (g/mol) 173.16
MDL Number MFCD00044734,MFCD00150724
InChI Key FEPBITJSIHRMRT-UHFFFAOYSA-M
Synonym sodium 4-hydroxybenzenesulfonate dihydrate, unii-5nh81n759q, phenol-4-sulfonic acid sodium salt dihydrate, 4-hydroxybenzenesulfonic acid sodium salt dihydrate, benzenesulfonic acid, 4-hydroxy-, monosodium salt, dihydrate, 4-hydroxybenzenesulfonic acid sodium salt, sodium 4-hydroxybenzene-1-sulfonate dihydrate, phenolsulphonate sodium usp, acmc-2098hs, sodium p-phenolsulfonate dihydrate
PubChem CID 23666329
IUPAC Name 4-hydroxybenzene-1-sulfonate
SMILES OC1=CC=C(C=C1)S([O-])(=O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
S082425G
View Documents
TCI America
S082425G
25 g
Each for $33.56
Only null left
 
S0824500G
View Documents
TCI America
S0824500G
500 g
Each for $160.30
Only null left
 
Description

Description

Specifications

Chemical Identifiers

10580-19-5
173.16
FEPBITJSIHRMRT-UHFFFAOYSA-M
23666329
OC1=CC=C(C=C1)S([O-])(=O)=O
C6H5O4S
MFCD00044734,MFCD00150724
sodium 4-hydroxybenzenesulfonate dihydrate, unii-5nh81n759q, phenol-4-sulfonic acid sodium salt dihydrate, 4-hydroxybenzenesulfonic acid sodium salt dihydrate, benzenesulfonic acid, 4-hydroxy-, monosodium salt, dihydrate, 4-hydroxybenzenesulfonic acid sodium salt, sodium 4-hydroxybenzene-1-sulfonate dihydrate, phenolsulphonate sodium usp, acmc-2098hs, sodium p-phenolsulfonate dihydrate
4-hydroxybenzene-1-sulfonate

Specifications

10580-19-5
C6H5O4S
25 g
FEPBITJSIHRMRT-UHFFFAOYSA-M
4-hydroxybenzene-1-sulfonate
23666329
≥98.0% (HPLC,T)
Sodium 4-Hydroxybenzenesulfonate Dihydrate
White-Yellow
MFCD00044734,MFCD00150724
sodium 4-hydroxybenzenesulfonate dihydrate, unii-5nh81n759q, phenol-4-sulfonic acid sodium salt dihydrate, 4-hydroxybenzenesulfonic acid sodium salt dihydrate, benzenesulfonic acid, 4-hydroxy-, monosodium salt, dihydrate, 4-hydroxybenzenesulfonic acid sodium salt, sodium 4-hydroxybenzene-1-sulfonate dihydrate, phenolsulphonate sodium usp, acmc-2098hs, sodium p-phenolsulfonate dihydrate
OC1=CC=C(C=C1)S([O-])(=O)=O
173.16
196.15
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-1958

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'