missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium 1-heptanesulfonate monohydrate, 99.0% (T), MilliporeSigma™ Supelco™
Sodium 1-heptanesulfonate monohydrate can be used as an ion-pairing (IP) reagent, to develop ion-pair reversed phase liquid chromatography method, for the determination of antituberculosis drug ethambutol hydrochloride.
$142.65 - $271.38
Chemical Identifiers
CAS | 207300-90-1 |
---|---|
Molecular Formula | C7H17NaO4S |
Molecular Weight (g/mol) | 220.26 |
MDL Number | MFCD00149550 |
InChI Key | XWZCREJRXRKIRQ-UHFFFAOYSA-M |
Synonym | 1-Heptanesulfonic acid sodium salt monohydrate |
IUPAC Name | sodium heptane-1-sulfonate hydrate |
SMILES | O.[Na+].CCCCCCCS([O-])(=O)=O |
Chemical Identifiers
207300-90-1 | |
220.26 | |
XWZCREJRXRKIRQ-UHFFFAOYSA-M | |
sodium heptane-1-sulfonate hydrate |
C7H17NaO4S | |
MFCD00149550 | |
1-Heptanesulfonic acid sodium salt monohydrate | |
O.[Na+].CCCCCCCS([O-])(=O)=O |
Specifications
207300-90-1 | |
CH3(CH2)5 CH2SO3Na · H2O | |
5 g | |
XWZCREJRXRKIRQ-UHFFFAOYSA-M | |
sodium heptane-1-sulfonate hydrate | |
220.26 | |
For ion pair chromatography |
C7H17NaO4S | |
MFCD00149550 | |
1-Heptanesulfonic acid sodium salt monohydrate | |
O.[Na+].CCCCCCCS([O-])(=O)=O | |
220.26 | |
≥99.0% (T) |