missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Keystone Safety White SMS Disposable Anti-Static Frocks
Frocks constructed of SMS composite fabric feature knit wrists, elastic thumb loops, and three pockets. They provide particulate and liquid barrier suitable for ISO Class 6 cleanrooms.
$223.70 - $243.47
Specifications
Product Type | Controlled Environments Frock |
---|---|
Material | SMS |
Color | White |
Disposable | Single-use |
Sterility | Non-sterile |
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
19-055-476
|
Keystone Safety
CEF3WKSMSSMWHITE |
Small |
Case of 30 Each for $223.70
|
|
|||||
19-055-478
|
Keystone Safety
CEF3WKSMSLRGWHITE |
Large |
Case of 30 Each for $223.70
|
|
|||||
19-055-479
|
Keystone Safety
CEF3WKSMSXLWHITE |
X-Large |
Case of 30 Each for $223.70
|
|
|||||
19-055-480
|
Keystone Safety
CEF3WKSMS2XLWHITE |
2X-Large |
Case of 30 Each for $223.70
|
|
|||||
19-055-482
|
Keystone Safety
CEF3WKSMS3XLWHITE |
3X-Large |
Case of 30 Each for $233.57
|
|
|||||
19-055-483
|
Keystone Safety
CEF3WKSMS4XLWHITE |
4X-Large |
Case of 30 Each for $243.47
|
|
|||||
Description
- Made from SMS composite fabric
- Disposable
- Highly breathable and comfortable
- ISO Class 6 cleanroom compatible
- Low shedding and anti-static to protect sensitive products
- Provide particulate barrier and liquid repellant
- Ideal for food processing, laboratories, biohazards, medical, metal surface treating, pharmaceutical
- Knit cuffs with elastic thumb loops, three pockets, six snap closure, knit collar, extra-long knee-length, hanger loop
Specifications
Controlled Environments Frock | |
White | |
Non-sterile | |
Knit, Elastic Thumb Loop | |
Manufacturing, Food Service, Industrial, Medical, Pharmaceutical | |
Yes | |
Long Sleeve | |
Class 1000 (ISO 6) |
SMS | |
Single-use | |
Snaps | |
Serged | |
Knit | |
3 | |
10/Bag, 3 Bags/Lined Carton |