missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Wanzl SmartCell™ Shelving Pedestal Frame Hang Bar
Pedestal Frame Hang Bar for SmartCell™ Shelving.
$24.74 - $46.74
Specifications
For Use With (Application) | Shelving |
---|---|
For Use With (Equipment) | SmartCell™ |
Includes | 1 Hang Bar |
Color | Chrome |
Material | Steel |
Description
- Pedestal Frame Hang Bar
- Replacement part for SmartCell™ Shelving
Specifications
Shelving | |
1 Hang Bar | |
Steel |
SmartCell™ | |
Chrome |