missing translation for 'onlineSavingsMsg'
Learn More

Wanzl SmartCell™ Angled Basket

All fine mesh wire baskets are plated in hospital-grade chrome and the framework is finished with industrial strength powder coat to enhance the durability and longevity of the system.

$317.00 - $521.51

Specifications

Color Chrome
Material Steel
For Use With (Application) Shelving
Shelf Type Angled Basket
View More Specs

Products 6
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity  
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity  
11222056
View Documents
Wanzl
THPM1836BSKAG
18 x 36 in.
Each for $317.00
Only null left
 
11222057
View Documents
Wanzl
THPM1848BSKAG
18 x 48 in.
Each for $387.64
Only null left
 
11222058
View Documents
Wanzl
THPM1860BSKAG
18 x 60 in.
Each for $443.43
Only null left
 
11222059
View Documents
Wanzl
THPM2436BSKAG
24 x 36 in.
Each for $390.43
Only null left
 
11222060
View Documents
Wanzl
THPM2448BSKAG
24 x 48 in.
Each for $450.85
Only null left
 
11222061
View Documents
Wanzl
THPM2460BSKAG
24 x 60 in.
Each for $521.51
Only null left
 
Description

Description

  • Baskets are available in a flat configuration or can be angled for easier access.
  • Adjustable dividers can be added to customize
Specifications

Specifications

Chrome
Steel
Shelving
Angled Basket
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'