Learn More
United Scientific™ Slide Mailer With Cap, PP, Neutral DFS Item
Convenient way to transfer and store slides while keeping them separated.
$100.77 - $109.50
Specifications
Type Slide Mailer
Material Polypropylene
View More Specs
Products 2
Catalog Number Mfr. No. Slide Capacity Price Quantity  
Catalog Number Mfr. No. Slide Capacity Price Quantity  
S012575 DFS Item
View Documents
United Scientific™
62605
3
Pack of 100 Each for $109.50
Only null left
S012574 DFS Item
View Documents
United Scientific™
62606
5
Pack of 100 Each for $100.77
Only null left
Description
Description
  • A convenient and safe solution for holding and transferring microscope slides. The inside walls feature slotted channels to keep slides separated and to make removal easier. The lids close securely to keep the slides safe.
  • Accommodates microscope slides in two sizes:
    • 75.0 mm x 25.0 mm x (0.8 to 1.2 mm)
    • 76.0 mm x 26.0 mm x (0.8 to 1.2 mm)
  • Securely hold slides: 3-place option with screw cap, and 5-place option with hinge cap
  • Durable Polypropylene
  • Holds two common slide sizes
Specifications
Specifications
Slide Mailer
Polypropylene
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'