Learn More
Research Products Slide Mailer for 5 Slides, Plastic, Assorted, 25/cs DFS Item
Supplier:  Research Products Corporation 212620A
Mailers offer secure protection for storage, transportation or staining of microscope slides. Durable polypropylene container holds up to five standard 1 x 3 inch slides. Slotted interior channels keep slides separated and facilitate quick removal. Lid closes securely to keep slides safe.
Catalog No. 501978509
Only null left
Explore available promotions