Learn More
Eisco™ Slide Cabinets
Convenient storage for up to 5000 slides.
Supplier: Eisco™ BI0123D
Description
- Wooden cabinet drawers fitted with wooden grooves that hold slides vertically
- For 3 × 1 in. (75 × 25mm) slides
- Individual index cards and locks for each drawer
- Each drawer has index card holder
- With lock
Order Info
500 capacity cabinet holds five drawers for 100 slides each; 1000 capacity, 10 drawers for 100 slides each; 2000 capacity, 10 drawers for 200 slides each; and 5000 capacity, 25 drawers for 200 slides each.
Specifications
Slide Storage Cabinet | |
5000 | |
Lock |
5000 | |
For storing slides | |
Wood |