missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC51A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5139968
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: MEPGRTQIKL DPRYTADLLE VLKTNYGIPS ACFSQPPTAA QLLRALGP.
Essential component of the Ost-alpha/Ost-beta complex, a heterodimer that acts as the intestinal basolateral transporter responsible for bile acid export from enterocytes into portal blood. Efficiently transports the major species of bile acids (taurocholate). Taurine conjugates are transported more efficiently across the basolateral membrane than glycine-conjugated bile acids (By similarity). Can also transport steroids such as estrone 3-sulfate and dehydroepiandrosterone 3- sulfate, therefore playing a role in the enterohepatic circulation of sterols. Able to transport eicosanoids such as prostaglandin E2 (By similarity).Specifications
SLC51A | |
Polyclonal | |
Unconjugated | |
SLC51A | |
organic solute transporter alpha; organic solute transporter subunit alpha; organic solute transporter, alpha subunit; OSTA; OSTalpha; OST-alpha; SLC51A; solute carrier family 51 alpha subunit; solute carrier family 51 subunit alpha; solute carrier family 51, alpha subunit | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
200931 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q86UW1 | |
SLC51A | |
Recombinant protein corresponding to Human SLC51A. Recombinant protein control fragment (Product #RP-108854). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |