missing translation for 'onlineSavingsMsg'
Learn More

Simport™ Scientific Sample Tubes with External Threads - Graduated

Sample Tubes with External Threads - Graduated with white marking area.

Supplier:  Simport™ Scientific T5014TPR

 View more versions of this product

Catalog No. 36-103-0258


Only null left
Explore available promotions

Description

Description

  • Made of polypropylene
  • Smooth and uniform inner surface with reduced risk of contamination
  • Can be autoclaved at upto 121°C in upright position with caps loosened
  • Height of tube is with cap
  • Available in choice of self standing or with round bottom
  • Caps sold separately
  • Round bottom tubes can be can be centrifuged up to 17,000 g
Specifications

Specifications

Sample Tube
Autoclavable
Round
Storage and transportation of biological materials
75 mm
Non-sterile
0.13 oz.
12.5 mm
4 mL
Polypropylene
External thread, graduations and white writing area
Round Bottom
Polyethylene
Screw Cap
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions