missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver nitrate solution, Volumetric, Reag. Ph. Eur., 0.1Â M AgNO3 (0.1N), Honeywell Fluka™
Volumetric, Reag. Ph. Eur., 0.1Â M AgNO3 (0.1N)
Supplier: Honeywell Chemicals 3537525L
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
Specifications
Silver nitrate solution | |
2.5 L | |
MFCD00003414 | |
silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol | |
[Ag+].[O-][N+]([O-])=O | |
169.87 | |
CHEBI:32130 | |
0.1M |
7761-88-8 | |
AgNO3 | |
UN3082 | |
SQGYOTSLMSWVJD-UHFFFAOYSA-N | |
silver(1+) nitrate | |
24470 | |
169.87g/mol |
Chemical Identifiers
7761-88-8 | |
169.87 | |
SQGYOTSLMSWVJD-UHFFFAOYSA-N | |
24470 | |
[Ag+].[O-][N+]([O-])=O |
AgNO3 | |
MFCD00003414 | |
silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol | |
CHEBI:32130 |
Safety and Handling
P273-P280-P305 + P351 + P338-P337 + P313-P391-P501