missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver Nitrate, Crystal, USP, 99.8-100.5%, Spectrum™ Chemical
SI115, 7761-88-8, AgNO3
$3300.03 - $3300.03
Chemical Identifiers
CAS | 7761-88-8 |
---|---|
Molecular Formula | AgNO3 |
Molecular Weight (g/mol) | 169.87 |
MDL Number | MFCD00003414 |
InChI Key | SQGYOTSLMSWVJD-UHFFFAOYSA-N |
IUPAC Name | silver(1+) nitrate |
SMILES | [Ag+].[O-][N+]([O-])=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
18606236
|
Spectrum Chemical Mfg Cor
SI115100GM |
100 g | Amber Glass Bottle |
Each for $3,300.03
|
|
||||
Description
Spectrum™ Chemical Silver Nitrate, Crystal, USP is used as an antibiotic for indolent wounds, corn and callus removal and as a Gonorrheal ophthalmia neonatorum prophylaxis. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
7761-88-8 | |
169.87 | |
SQGYOTSLMSWVJD-UHFFFAOYSA-N | |
[Ag+].[O-][N+]([O-])=O |
AgNO3 | |
MFCD00003414 | |
silver(1+) nitrate |
Specifications
7761-88-8 | |
100 g | |
MFCD00003414 | |
[Ag+].[O-][N+]([O-])=O | |
169.87 | |
USP |
1 | |
AgNO3 | |
SQGYOTSLMSWVJD-UHFFFAOYSA-N | |
silver(1+) nitrate | |
99.8 to 100.5% | |
Amber Glass Bottle |