missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver(I) Trifluoromethanethiolate 95.0+%, TCI America™
$275.87 - $995.41
Chemical Identifiers
CAS | 811-68-7 |
---|---|
Molecular Formula | CHAgF3S |
Molecular Weight (g/mol) | 209.94 |
MDL Number | MFCD25563234 |
InChI Key | SVPQZGWGYCARGG-UHFFFAOYSA-N |
Synonym | Trifluoromethanethiol Silver(I) Salt |
PubChem CID | 24191737 |
IUPAC Name | trifluoromethanethiol silver |
SMILES | [Ag].FC(F)(F)S |
Chemical Identifiers
811-68-7 | |
209.94 | |
SVPQZGWGYCARGG-UHFFFAOYSA-N | |
24191737 | |
[Ag].FC(F)(F)S |
CHAgF3S | |
MFCD25563234 | |
Trifluoromethanethiol Silver(I) Salt | |
trifluoromethanethiol silver |
Specifications
811-68-7 | |
MFCD25563234 | |
Trifluoromethanethiol Silver(I) Salt | |
[Ag].FC(F)(F)S | |
209.94 | |
208.93 | |
Crystalline Powder |
CHAgF3S | |
1 g | |
SVPQZGWGYCARGG-UHFFFAOYSA-N | |
trifluoromethanethiol silver | |
24191737 | |
≥95.0% (T) | |
Silver(I) Trifluoromethanethiolate |
Safety and Handling
TSCA : No