missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver foil, 0.25mm (0.01in) thick, annealed, Premion™, 99.998% (metals basis)
CAS: 7440-22-4 | Ag | 107.87 g/mol
$230.34 - $2868.58
Chemical Identifiers
CAS | 7440-22-4 |
---|---|
Molecular Formula | Ag |
Molecular Weight (g/mol) | 107.87 |
MDL Number | MFCD00003397 |
InChI Key | BQCADISMDOOEFD-UHFFFAOYSA-N |
Synonym | argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber |
PubChem CID | 23954 |
ChEBI | CHEBI:9141 |
IUPAC Name | silver |
SMILES | [Ag] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA42474FI
|
Thermo Scientific Chemicals
042474FI |
50 x 50 mm |
Each for $230.34
|
|
|||||
AA42474GH
|
Thermo Scientific Chemicals
042474GH |
100 x 100 mm |
Each for $705.95
|
|
|||||
AA42474HP
|
Thermo Scientific Chemicals
042474HP |
100 x 500 mm |
Each for $2,868.58
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7440-22-4 | |
107.87 | |
BQCADISMDOOEFD-UHFFFAOYSA-N | |
23954 | |
silver |
Ag | |
MFCD00003397 | |
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber | |
CHEBI:9141 | |
[Ag] |
Specifications
7440-22-4 | |
Characteristic | |
Foil | |
MFCD00003397 | |
BQCADISMDOOEFD-UHFFFAOYSA-N | |
silver | |
23954 | |
99.998% | |
Silver foil, 0.25mm (0.01 in.) thick, Annealed, Temper: soft |
50 x 50 mm | |
≈6.6g/50x50mm | |
Ag | |
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber | |
[Ag] | |
107.87 | |
CHEBI:9141 | |
Premion™ | |
(metals basis) |
Safety and Handling
EINECSNumber : 231-131-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only