missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silica Gel, Honeywell Fluka™
With moisture indicator free of heavy metals
$203.48 - $3493.78
Chemical Identifiers
CAS | 112926-00-8 |
---|---|
Molecular Formula | O2Si |
Molecular Weight (g/mol) | 60.083 |
MDL Number | MFCD00011232 |
InChI Key | VYPSYNLAJGMNEJ-UHFFFAOYSA-N |
PubChem CID | 24261 |
ChEBI | CHEBI:30563 |
IUPAC Name | dioxosilane |
SMILES | O=[Si]=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
6002004
|
Honeywell-Fluka
13767500G |
500 g |
Each for $203.48
|
|
|||||
6002002
|
Honeywell Chemicals
137671KG |
1 kg |
Each for $340.45
|
|
|||||
6002003
|
Honeywell Chemicals
1376725KG |
2.5 kg |
Each for $674.70
|
|
|||||
60044822
|
Honeywell Chemicals
137676X500G |
6 x 500 g |
Case of 6 Each for $1,052.08
|
|
|||||
60044820
|
Honeywell-Fluka
137676X1KG |
6 x 1 kg |
Case of 6 Each for $1,759.25
|
|
|||||
60044821
|
Honeywell Chemicals
137676X25KG |
6 x 2.5 kg |
Case of 6 Each for $3,493.78
|
|
|||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
112926-00-8 | |
60.083 | |
VYPSYNLAJGMNEJ-UHFFFAOYSA-N | |
CHEBI:30563 | |
O=[Si]=O |
O2Si | |
MFCD00011232 | |
24261 | |
dioxosilane |
Specifications
112926-00-8 | |
500 g | |
MFCD00011232 | |
VYPSYNLAJGMNEJ-UHFFFAOYSA-N | |
dioxosilane | |
24261 | |
Silica Gel |
Silica Gel | |
O2Si | |
NONH for all modes of transport | |
O=[Si]=O | |
60.083 | |
CHEBI:30563 |
Safety and Handling
EINECSNumber : 231-545-4