missing translation for 'onlineSavingsMsg'
Learn More

Silica Gel, Grade 923, 100-200 Mesh, Spectrum™ Chemical

SIL40, 7631-86-9, SiO2

$2861.97 - $85966.67

Chemical Identifiers

CAS 7631-86-9
Molecular Formula O2Si
Molecular Weight (g/mol) 60.08
MDL Number MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
IUPAC Name silanedione
SMILES O=[Si]=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18606208
View Documents
Spectrum Chemical Mfg Cor
SIL40500GM
500 g Amber Glass Bottle
Each for $2,861.97
Only null left
 
18606209
View Documents
Spectrum Chemical Mfg Cor
SIL40500GMCS6
6 x 500 g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
18606210
View Documents
Spectrum Chemical Mfg Cor
SIL4025KGBL
25 kg Fiber Drum N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Spectrum™ Chemical Silica Gel, Grade 923, 100-200 Mesh is a porous and granular form of silicon dioxide which is synthetically made from sodium silicate. In chemistry, the silica gel is used as a stationary phase in chromatography. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Specifications

Chemical Identifiers

7631-86-9
60.08
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
O=[Si]=O
O2Si
MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
silanedione

Specifications

7631-86-9
500 g
MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
silanedione
Reagent
1
O2Si
Amber Glass Bottle
O=[Si]=O
60.08
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'