missing translation for 'onlineSavingsMsg'
Learn More

Watson Gloves Shock Trooper Lined Gauntlet

Shock Trooper Lined Gauntlet is made of lightweight 3M Thinsulate™ C40 (palm) and C100 (back) lining that keeps hands warm in cold conditions and provides better dexterity.

$415.34 - $415.34

Specifications

Back Material Full Grain Goatskin Leather
Certifications/Compliance ANSI CUT, ANSI Impact, ANSI Puncture
Certifications/Compliance (Europe) EN388
Chemotherapeutic Rated No
Chlorinated No
View More Specs

Products 5
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
17-099-756
View Documents
Watson Gloves
95785GS
Small
Case of 6 Pair for $415.34
Only null left
 
17-099-993
View Documents
Watson Gloves
95785GM
Medium
Case of 6 Pair for $415.34
Only null left
 
17-099-755
View Documents
Watson Gloves
95785GL
Large
Case of 6 Pair for $415.34
Only null left
 
17-099-743
View Documents
Watson Gloves
95785GX
X-Large
Case of 6 Pair for $415.34
Only null left
 
17-099-974
View Documents
Watson Gloves
95785GXXL
2X-Large
Case of 6 Pair for $415.34
Only null left
 
Description

Description

  • Lightweight 3M Thinsulate™ C40 (palm) and C100 (back) lining
  • Cutshield™ is a cut resistant liner made from a blend of P-aramid, steel and modacrylic fibres
  • Dryhide™ water and oil resistant full-grain goatskin leather palm and back of hand
  • Knuckle and finger components feature D3O™ iA
  • Vibration absorbing EVA padded palm patch and wrist patch
  • Stitched with Aramid Fiber thread
  • Snug-fitting shirred elastic wrist with gauntlet style cuff
  • Partial proceeds go toward Wounded Warriors Canada
Specifications

Specifications

Full Grain Goatskin Leather
EN388
No
Cut and Sewn
Full Sleeve
Industrial
Black, Brown
Smooth
Yes
Work Gloves
Gunn
No
Oil and Gas, LNG, Industrial, Heavy Mechanical, Pipeline, Winter
Black
1 Pair of Gloves
Thinsulate
95785G
ANSI/ISEA 105-16 Level 4
Shock Trooper
Straight
5 in.
13 cm
11.5 cm
0.30 kg
ANSI CUT, ANSI Impact, ANSI Puncture
No
Uncoated
Leather Gore
P-aramid Blend Glove
Uncoated
Reusable
Ambidextrous
Goatskin Leather
Non-sterile
EN388:2X43FP
Full
Men's
Gauntlet
No
Lined
Full Grain Goatskin Leather
Cut, Impact, Puncture, Abrasion, Oil, Water, Cold
4
12 in.
30 cm
4.5 in.
0.65 lb.
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'