Learn More
Epredia™ Accessories for Hyperclean TruAir™ Workstation
For use with Epredia™ Hyperclean Pathology Workstations.
Supplier:  Epredia™ 9990611
 View more versions of this product
Catalog No. 28-601-34
Only null left
Explore available promotions
Description
Description
Accessories:
  • Charcoal Filter
  • Fine particle Filters
  • Formaldehyde Neutralizing (Potassium Permanganate) Filter
Specifications
Specifications
Workstation Accessories
Fine Particle Filter
Epredia Hyperclean Pathology Workstations, Models 1, 20, 22, 36, 60
Hyperclean™
Product Suggestions
Product Suggestions