Learn More
Epredia™ Accessories for Hyperclean TruAir™ Workstation
For use with Epredia™ Hyperclean Pathology Workstations.
Supplier: Epredia™ 9990615
Description
Accessories:- Charcoal Filter
- Fine particle Filters
- Formaldehyde Neutralizing (Potassium Permanganate) Filter
Specifications
Workstation Accessories | |
Size: 6.7 x 27.9 x 27.9cm | |
Epredia™ Hyperclean Pathology Workstations, model 2 | |
Hyperclean™ |