Learn More
MilliporeSigma™ Supelco™ SGE Plunger-Protected Syringe, Fixed Needle
Extended protection tube from top of the flange to help prevent plunger bending during injection
$401.88 - $401.88
Specifications
Needle Gauge | 26 G |
---|---|
Product Type | Plunger-Protection Syringe |
Needle Point Number | 2 |
Volume (Metric) | 10 μL |
Length (Metric) Needle | 50 mm |
Specifications
26 G | |
2 | |
50 mm | |
Fixed |
Plunger-Protection Syringe | |
10 μL | |
Bevel |