missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Selectophore™ Hydrogen ionophore I, Function Tested, MilliporeSigma™ Supelco™
Visit our Sensor Applications portal to learn more.
$220.20 - $713.08
Chemical Identifiers
MDL Number | MFCD00008971 |
---|---|
Synonym | Tridodecylamine; Proton ionophore I; Hydrogen ionophore I |
Chemical Identifiers
MFCD00008971 | |
Tridodecylamine; Proton ionophore I; Hydrogen ionophore I |
Specifications
102-87-4 | |
220°C to 228°C/0.03 mmHg | |
Bottomless Glass Bottle with Inserted Fused Cone | |
MFCD00008971 | |
Tridodecylamine; Proton ionophore I; Hydrogen ionophore I | |
521.99 | |
Selectophore |
0.823 g/mL (at 25°C) | |
113°C | |
[CH3(CH2)11]3 N | |
100 mg | |
>1 (vs. air) | |
Selectophore |
Selectophore is a trademark of Sigma-Aldrich Chemie GmbH