missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Selectophore™ Dipentyl phthalate, 99.0%, MilliporeSigma™ Supelco™
Plasticizer for PVC membrane electrodes
$142.39 - $476.52
Chemical Identifiers
CAS | 131-18-0 |
---|---|
Molecular Formula | C18H26O4 |
Molecular Weight (g/mol) | 306.40 |
MDL Number | MFCD00041934 |
InChI Key | IPKKHRVROFYTEK-UHFFFAOYSA-N |
IUPAC Name | 1,2-dipentyl benzene-1,2-dicarboxylate |
SMILES | CCCCCOC(=O)C1=CC=CC=C1C(=O)OCCCCC |
Chemical Identifiers
131-18-0 | |
306.40 | |
IPKKHRVROFYTEK-UHFFFAOYSA-N | |
CCCCCOC(=O)C1=CC=CC=C1C(=O)OCCCCC |
C18H26O4 | |
MFCD00041934 | |
1,2-dipentyl benzene-1,2-dicarboxylate |
Specifications
131-18-0 | |
C18H26O4 | |
n20/D 1.490 | |
1 mL | |
CCCCCOC(=O)C1=CC=CC=C1C(=O)OCCCCC | |
306.40 | |
≥99.0% (GC) | |
Selectophore |
1.025 g/mL (at 20°C) | |
C18H26O4 | |
MFCD00041934 | |
IPKKHRVROFYTEK-UHFFFAOYSA-N | |
1,2-dipentyl benzene-1,2-dicarboxylate | |
306.40 | |
Selectophore™ |
Safety and Handling
RTECSNumber : TI1930000
Selectophore is a trademark of Sigma-Aldrich Chemie GmbH