missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Selectophore™ Diisodecyl phthalate, ≥99.0%, MilliporeSigma™ Supelco™
Diisodecyl phthalate (DIDP) belongs to phthalate group, and is a dialkyl or aryl/alkyl diesters of phthalic acid. It is mostly used as a plasticizer in manufacturing polymers and other consumer products.
$114.67 - $466.68
Specifications
CAS | 26761-40-0 |
---|---|
Density | 0.965 g/mL (at 20°C) |
Flash Point | 275°C |
Linear Formula | C28H46O4 |
Quantity | 10 mL |
Formula Weight | 446.66 |
Percent Purity | ≥99.0% (sum of isomers, GC); ≥99.0% |
Grade | Selectophore |
Product Line | Selectophore |
Specifications
26761-40-0 | |
275°C | |
10 mL | |
≥99.0% (sum of isomers, GC); ≥99.0% | |
Selectophore |
0.965 g/mL (at 20°C) | |
C28H46O4 | |
446.66 | |
Selectophore |
Safety and Handling
RTECSNumber : TI1270000
Selectophore is a trademark of Sigma-Aldrich Chemie GmbH