missing translation for 'onlineSavingsMsg'
Learn More

Savory Oil, Summer Variety, Spectrum™ Chemical

$330.27 - $2084.73

Chemical Identifiers

CAS 8016-68-0
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18613080
View Documents
Spectrum Chemical Mfg Cor
S109625ML
25 mL Amber Glass Bottle
Each for $330.27
Only null left
 
18613081
View Documents
Spectrum Chemical Mfg Cor
S1096100ML
100 mL Amber Glass Bottle
Each for $883.50
Only null left
 
18613082
View Documents
Spectrum Chemical Mfg Cor
S1096500ML
500 mL Amber Glass Bottle
Each for $2,084.73
Only null left
 
Description

Description

Specifications

Chemical Identifiers

8016-68-0

Specifications

8016-68-0
Amber Glass Bottle
Reagent
1
25 mL
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.