missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Salicylic Acid, Powder, USP, 98-102%, Spectrum™ Chemical
SA130, 69-72-7, C7H6O3
Supplier: Spectrum Chemical Mfg Cor SA13025KGBL
Description
Spectrum™ Chemical Salicylic Acid, Powder, USP is a topical application used to treat skin conditions that involve scaling or overgrowth of skin cells such as psoriasis, dandruff, corns, calluses, and warts. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
69-72-7 | |
C7H6O3 | |
YGSDEFSMJLZEOE-UHFFFAOYSA-N | |
2-hydroxybenzoic acid | |
98 to 102% | |
25 kg | |
0.005 |
1 | |
MFCD00002439 | |
OC(=O)C1=CC=CC=C1O | |
138.12 | |
USP | |
0.0005 | |
Fiber Drum |
Chemical Identifiers
69-72-7 | |
138.12 | |
YGSDEFSMJLZEOE-UHFFFAOYSA-N | |
OC(=O)C1=CC=CC=C1O |
C7H6O3 | |
MFCD00002439 | |
2-hydroxybenzoic acid |