missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Salicylaldehyde, 98%, Spectrum™ Chemical
SA122, 90-02-8, C7H6O2
$859.97 - $39362.11
Chemical Identifiers
CAS | 90-02-8 |
---|---|
Molecular Formula | C7H6O2 |
Molecular Weight (g/mol) | 122.12 |
MDL Number | MFCD00003317 |
InChI Key | SMQUZDBALVYZAC-UHFFFAOYSA-N |
IUPAC Name | 2-hydroxybenzaldehyde |
SMILES | OC1=CC=CC=C1C=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
18606141
|
Spectrum Chemical Mfg Cor
SA1221LT |
1 L | Amber Glass Bottle |
Each for $859.97
|
|
||||
18606142
|
Spectrum Chemical Mfg Cor
SA122200LTBL |
200 L | Drum |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Spectrum™ Chemical Salicylaldehyde , also known as Salicylic aldehyde or 2-Hydroxybenzaldehyde, is a precursor to a several types of commercially important chelating agents. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.Chemical Identifiers
90-02-8 | |
122.12 | |
SMQUZDBALVYZAC-UHFFFAOYSA-N | |
OC1=CC=CC=C1C=O |
C7H6O2 | |
MFCD00003317 | |
2-hydroxybenzaldehyde |
Specifications
90-02-8 | |
Amber Glass Bottle | |
1.571-1.575 | |
1 L | |
OC1=CC=CC=C1C=O | |
122.12 | |
Reagent |
1 | |
C7H6O2 | |
MFCD00003317 | |
SMQUZDBALVYZAC-UHFFFAOYSA-N | |
2-hydroxybenzaldehyde | |
98% |