Learn More
Slice™ Safety Cutter
Green cutter with an ambidextrous design
Supplier:  Slice™ DTC2100100
Catalog No. 29-636-2082
Only null left
Explore available promotions
Description
Description
  • Micro-ceramic blade type
  • Rubberized non-slip finish
  • Non-sparking, non-conductive blade
  • Chemically inert blade that never rusts
  • Built in magnet and key-ring hole
  • Reduces injuries
  • Lowers cost
Specifications
Specifications
Slice™ Safety Cutter
Ceramic
Product Suggestions
Product Suggestions