missing translation for 'onlineSavingsMsg'
Learn More

Medicom™ SafeTouch™ Rejuvenate™ Latex Medical Examination Glove with Textured Fingertips

Medicom™ SafeTouch™ Rejuvenate™ Latex Medical Examination Gloves with Textured Fingertips, Infused with lanolin and vitamin E.

$23.13 - $25.32

Specifications

Chlorinated No
Accelerator Free No
Applications Healthcare
AQL 1.5
Color Cream
View More Specs

Products 5
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
29-635-7739
View Documents
Medicom™
1163A
X-Small
Pack of 100 Each for $23.13
Case of 10 Pack for $231.30
Only null left
 
29-635-7740
View Documents
Medicom™
1163B
Small
Pack of 100 Each for $24.39
Case of 10 Pack for $243.90
Only null left
 
29-635-7741
Accessories View Documents
Medicom™
1163C
Medium
Pack of 100 Each for $25.32
Case of 10 Pack for $253.20
Only null left
 
29-635-7742
Accessories View Documents
Medicom™
1163D
Large
Pack of 100 Each for $24.78
Case of 10 Pack for $247.80
Only null left
 
29-635-7743
Accessories View Documents
Medicom™
1163E
X-Large
Pack of 100 Each for $23.32
Case of 10 Pack for $233.20
Only null left
 
Description

Description

  • Heavier weight, highly resistant glove.
  • Made from natural rubber latex for an ergonomic fit and optimal tear, puncture and chemical resistance.
  • Textured fingertips for a secure grip, even when wet.
  • Skin-friendly: Infused with lanolin and vitamin E to moisturize and soften hands.
  • 1000/Cs.
Specifications

Specifications

No
Healthcare
Cream
Fully Textured
Powder Free
No
No
1.5
Single-use
No
Non-sterile
Beaded
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'