missing translation for 'onlineSavingsMsg'
Learn More

Medicom™ SafeTouch™ Connect™ Latex Medical Examination Gloves

SafeTouch™ Connect™ Latex Medical Examination Gloves - Low protein content.

$17.69 - $20.17

Specifications

Accelerator Free No
Applications Healthcare
AQL 1.5
Color Cream
Disposable Single-use
View More Specs

Products 5
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
36-100-4629
View Documents
Medicom™
1124A
X-Small
Pack of 100 Each for $20.17
Case of 10 Pack for $201.70
Only null left
 
36-100-4630
View Documents
Medicom™
1124B
Small
Pack of 100 Each for $18.52
Case of 10 Pack for $185.20
Only null left
 
36-100-4631
View Documents
Medicom™
1124C
Medium
Pack of 100 Each for $19.98
Case of 10 Pack for $199.80
Only null left
 
36-100-4632
View Documents
Medicom™
1124D
Large
Pack of 100 Each for $20.13
Case of 10 Pack for $201.30
Only null left
 
36-100-4633
View Documents
Medicom™
1124E
X-Large
Pack of 100 Each for $17.69
Case of 10 Pack for $176.90
Only null left
 
Description

Description

  • Heavier weight, highly resistant, low protein content latex glove.
  • Fully textured for optimal grip.
  • 1000/Cs.
Specifications

Specifications

No
1.5
Single-use
No
Powder Free
No
Healthcare
Cream
Fully Textured
Latex
Non-sterile
Beaded
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'