Learn More
Medicom™ SafeMask Tailormade Earloop Mask
SafeMask Tailormade Earloop Face Masks are available in 50/box, 10 boxes/Cs. and 500/Cs.
Supplier: Medicom™ 2072
Description
- Dual-fit nose and chin pieces adjust to form a tight seal around the face to reduce gapping and ensure a personalized fit.
- Soft non-woven material enhances breathability.
- Smooth stretchy latex-free ultrasonically sealed earloops prevent irritation and allow easy removal with a lower risk of self-contamination.
Specifications
Medical/Procedural Face Mask | |
Blue | |
Non-sterile | |
Single-use |
ASTM Level 3 | |
Earloops | |
Pleated | |
Yes |